Learn More
Medchemexpress LLC Cromolyn disodium | 15826-37-6 | 100.0% | 1 ML
SDP
Catalog No. 5000406693
Encompass_Preferred
Cromolyn disodium (Cromoglycate) is an orally active GSK-3β inhibitor with an IC50 of 2.0 μM. It functions as a mast cell stabilizer by inhibiting the release of mediators from mast cells. This action helps to regulate reflex bronchoconstriction and reduce non-specific bronchial hyperreactivity, making it suitable for research into bronchial asthma.
  • Exhibits anti-inflammatory effects
  • Exhibits anti-allergic effects
  • Exhibits anti-histamine effects
  • Exhibits anti-cancer effects
  • Exhibits neuroprotective effects
Catalog No. 50-004-06693 Supplier Medchemexpress LLC Supplier No. HYB0320A10MM/1ML
Cromolyn disodium (Cromoglycate) is an orally active GSK-3β inhibitor with an IC50 of 2.0 μM. It functions as a mast cell stabilizer by inhibiting the release of mediators from mast cells. This action helps to regulate reflex bronchoconstriction and reduce non-specific bronchial hyperreactivity, making it suitable for research into bronchial asthma.
  • Exhibits anti-inflammatory effects
  • Exhibits anti-allergic effects
  • Exhibits anti-histamine effects
  • Exhibits anti-cancer effects
  • Exhibits neuroprotective effects