Learn More
Medchemexpress LLC Cromolyn (disodium) | 15826-37-6 | 89.2% | 100 MG
SDP
Catalog No. 5000406715
Encompass_Preferred
Cromolyn disodium is an analytical standard for research applications. It acts as an orally active GSK-3β inhibitor and a mast cell stabilizer, inhibiting mediator release, regulating bronchoconstriction, and reducing bronchial hyperreactivity. Used in asthma research, it also shows anti-inflammatory, anti-allergic, anti-histamine, anti-cancer, and neuroprotective effects.
  • Analytical standard for research
  • GSK-3β inhibitor and mast cell stabilizer
  • Inhibits mediator release
  • Regulates bronchoconstriction
  • Reduces bronchial hyperreactivity
  • Applied in asthma research
  • Anti-inflammatory, anti-allergic, anti-histamine, anti-cancer, and neuroprotective effects
  • Suitable for HPLC, GC, and MS
  • White to off-white solid
  • Stable under recommended storage
Catalog No. 50-004-06715 Supplier Medchemexpress LLC Supplier No. HYB0320AR100MG
Cromolyn disodium is an analytical standard for research applications. It acts as an orally active GSK-3β inhibitor and a mast cell stabilizer, inhibiting mediator release, regulating bronchoconstriction, and reducing bronchial hyperreactivity. Used in asthma research, it also shows anti-inflammatory, anti-allergic, anti-histamine, anti-cancer, and neuroprotective effects.
  • Analytical standard for research
  • GSK-3β inhibitor and mast cell stabilizer
  • Inhibits mediator release
  • Regulates bronchoconstriction
  • Reduces bronchial hyperreactivity
  • Applied in asthma research
  • Anti-inflammatory, anti-allergic, anti-histamine, anti-cancer, and neuroprotective effects
  • Suitable for HPLC, GC, and MS
  • White to off-white solid
  • Stable under recommended storage