Learn More
Medchemexpress LLC Cromolyn disodium | 15826-37-6 | 99.98% | 5 G
SDP
Catalog No. 5000406342
Encompass_Preferred
Cromolyn disodium is an orally active GSK-3β inhibitor and a mast cell stabilizer. It inhibits the release of mediators from mast cells, regulates reflex bronchoconstriction, and reduces non-specific bronchial hyperreactivity. It is used in the research of bronchial asthma and exhibits anti-inflammatory, anti-allergic, anti-histamine, anti-cancer, and neuroprotective effects.
  • Orally active GSK-3β inhibitor
  • Mast cell stabilizer
  • Inhibits mediator release from mast cells
  • Regulates reflex bronchoconstriction
  • Reduces non-specific bronchial hyperreactivity
  • Anti-inflammatory properties
  • Anti-allergic effects
  • Anti-histamine activity
  • Anti-cancer properties
  • Neuroprotective effects
Catalog No. 50-004-06342 Supplier Medchemexpress LLC Supplier No. HYB0320A5G
Cromolyn disodium is an orally active GSK-3β inhibitor and a mast cell stabilizer. It inhibits the release of mediators from mast cells, regulates reflex bronchoconstriction, and reduces non-specific bronchial hyperreactivity. It is used in the research of bronchial asthma and exhibits anti-inflammatory, anti-allergic, anti-histamine, anti-cancer, and neuroprotective effects.
  • Orally active GSK-3β inhibitor
  • Mast cell stabilizer
  • Inhibits mediator release from mast cells
  • Regulates reflex bronchoconstriction
  • Reduces non-specific bronchial hyperreactivity
  • Anti-inflammatory properties
  • Anti-allergic effects
  • Anti-histamine activity
  • Anti-cancer properties
  • Neuroprotective effects