Learn More
Apexbio Technology LLC Cromolyn sodium 15826-37-6 1g
SDP
Catalog No. 5000094213
Encompass_Preferred
Cromolyn sodium (CAS 15826-37-6) is a small molecule that functions as a mast cell membrane stabilizer It inhibits antigen-induced histamine release by preventing degranulation of sensitized mast cells Due to minimal gastrointestinal absorption cromolyn sodium is typically administered via inhalation in powder form Preclinical studies have demonstrated that cromolyn sodium suppresses both immediate and late-phase asthmatic responses and reduces the release of non-histamine chemical mediators Additionally it prevents ciliary transport impairment associated with bronchial provocation and can mitigate symptoms in exercise-induced asthma models These properties make cromolyn sodium a useful tool for investigating allergic and inflammatory mechanisms in respiratory research
Catalog No. 50-000-94213 Supplier Apexbio Technology LLC Supplier No. B33001000
Cromolyn sodium (CAS 15826-37-6) is a small molecule that functions as a mast cell membrane stabilizer It inhibits antigen-induced histamine release by preventing degranulation of sensitized mast cells Due to minimal gastrointestinal absorption cromolyn sodium is typically administered via inhalation in powder form Preclinical studies have demonstrated that cromolyn sodium suppresses both immediate and late-phase asthmatic responses and reduces the release of non-histamine chemical mediators Additionally it prevents ciliary transport impairment associated with bronchial provocation and can mitigate symptoms in exercise-induced asthma models These properties make cromolyn sodium a useful tool for investigating allergic and inflammatory mechanisms in respiratory research