Learn More
Medchemexpress LLC HY-B0320A 500mg Medchemexpress, Cromolyn (sodium) CAS:15826-37-6 Purity:>98%
SDP
Catalog No. 502082466
Encompass_Preferred
Medchemexpress, HY-B0320A 500mg Cromolyn (sodium) CAS:15826-37-6 Cromolyn sodium (Disodium Cromoglycate; FPL-670) is an antiallergic drug. Cromolyn sodium is a GSK-3β inhibitor with an IC50 of 2.0 µM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2466 Supplier Medchemexpress LLC Supplier No. HYB0320A500MG
Medchemexpress, HY-B0320A 500mg Cromolyn (sodium) CAS:15826-37-6 Cromolyn sodium (Disodium Cromoglycate; FPL-670) is an antiallergic drug. Cromolyn sodium is a GSK-3β inhibitor with an IC50 of 2.0 µM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.