Learn More
Cayman Chemical CromolynsodIum salt hydrAT 25g
SDP
Catalog No. 502981133
Encompass_Preferred
A chromone complex that acts as a mast cell stabilizer to prevent the release of histamine
Catalog No. 50-298-1133 Supplier Cayman Chemical Supplier No. 2137925 G
A chromone complex that acts as a mast cell stabilizer to prevent the release of histamine