Learn More
ATCC Cronobacter sakazakii; Strain CDC 4562-70
Catalog No. 502383542
Encompass
Cronobacter sakazakii strain CDC 4562-70 was isolated from the throat of a human child. This whole-genome sequenced bacterial type strain can be used in food testing.
Catalog No. 50-238-3542 Supplier ATCC Supplier No. 29544
 Restricted Item
Add to Cart
Add to Cart
Cronobacter sakazakii strain CDC 4562-70 was isolated from the throat of a human child. This whole-genome sequenced bacterial type strain can be used in food testing.