Learn More
NEOGEN Cronobacter Selective Broth, 500 g
SDP
Catalog No. 502015789
Encompass_Preferred
Cronobacter Selective Broth (CSB) is recommended by ISO 22964 for the isolation and identification of Cronobacter spp. (formerly Enterobacter sakazakii), and is not intended for use in the diagnosis of disease or other conditions in humans.Although rarely causing infections in immunocompetent adults, members of the Cronobacter genus have been implicated in sepsis, meningitis and necrotizing enterocolitis with a high death rate in neonates. These opportunistic pathogens are common in the environment and their ability to survive desiccation presents a significant risk for post-pasteurization contamination and survival in spray dried milk products. Enzymatic digest of animal tissues and meat extract provide a source of nitrogen and essential vitamins.For more information visit https://www.neogen.com/solutions/Microbiology/cronobacter-selective-broth
Catalog No. 50-201-5789 Supplier NEOGEN Supplier No. 700004670
Cronobacter Selective Broth (CSB) is recommended by ISO 22964 for the isolation and identification of Cronobacter spp. (formerly Enterobacter sakazakii), and is not intended for use in the diagnosis of disease or other conditions in humans.Although rarely causing infections in immunocompetent adults, members of the Cronobacter genus have been implicated in sepsis, meningitis and necrotizing enterocolitis with a high death rate in neonates. These opportunistic pathogens are common in the environment and their ability to survive desiccation presents a significant risk for post-pasteurization contamination and survival in spray dried milk products. Enzymatic digest of animal tissues and meat extract provide a source of nitrogen and essential vitamins.For more information visit https://www.neogen.com/solutions/Microbiology/cronobacter-selective-broth