Learn More
Cayman Chemical CropropamIde 10mg
SDP
Catalog No. 502991403
Encompass_Preferred
A component of prethcamide; increases locomotor activity in rats at 33.69 and 59.97 mg/kg
Catalog No. 50-299-1403 Supplier Cayman Chemical Supplier No. 3088810 MG
A component of prethcamide; increases locomotor activity in rats at 33.69 and 59.97 mg/kg