Learn More
MSC Crosby Grab Hook, 3/8 in, Grade 100, Alloy Steel, 8800 lb WLL
Catalog No. 5000535173
Encompass
This grab hook is designed for demanding lifting applications, featuring an 8800 lb working load limit. Constructed from Grade 100 alloy steel, it offers exceptional strength and durability. The clevis attachment allows direct connection to chain slings for secure overhead lifting.
  • Clevis pin attachment for direct connection to chain slings.
  • Slim profile for tight rigging applications.
  • Forged alloy steel construction provides superior strength.
  • Individually proof tested at 2-1/2 times the working load limit.
  • Fatigue rated for 20,000 cycles.
  • Ultimate load capacity is 4 times the working load limit.
  • Traceable with product identification markings.
  • Rated for overhead lifting.
  • Not suitable as a shortening hook.
  • Operating temperature range from -40 to 392 degrees F.
Catalog No. 50-005-35173 Supplier MSC Supplier No. 53595435
This grab hook is designed for demanding lifting applications, featuring an 8800 lb working load limit. Constructed from Grade 100 alloy steel, it offers exceptional strength and durability. The clevis attachment allows direct connection to chain slings for secure overhead lifting.
  • Clevis pin attachment for direct connection to chain slings.
  • Slim profile for tight rigging applications.
  • Forged alloy steel construction provides superior strength.
  • Individually proof tested at 2-1/2 times the working load limit.
  • Fatigue rated for 20,000 cycles.
  • Ultimate load capacity is 4 times the working load limit.
  • Traceable with product identification markings.
  • Rated for overhead lifting.
  • Not suitable as a shortening hook.
  • Operating temperature range from -40 to 392 degrees F.