Learn More
MSC Crosby Screw Pin Anchor Shackle, 1/2 in, 2 t WLL, G-209
Catalog No. 5000515494
Encompass
This screw pin anchor shackle is forged, quenched, and tempered, featuring alloy pins for durability. It is fatigue-rated and designed for a wide range of temperatures, from -40° C to 204° C. The shackle meets or exceeds all requirements of ASME B30.26 and federal specifications, ensuring reliable performance in demanding applications. Each shackle is permanently marked with its working load limit and grade '6', signifying genuine quality.
  • Capacities from 1/3 through 55 metric tons.
  • Sizes 7/16 inch and above are hot dip galvanized.
  • Fatigue rated design ensures long-term performance.
  • Proof testing with certificates can be furnished upon request.
  • Can meet charpy requirements of 42 Joules average at -20° C upon special request.
  • Maximum proof load is 2 times the working load limit.
  • Minimum ultimate strength is 6 times the working load limit.
Catalog No. 50-005-15494 Supplier MSC Supplier No. 81008633
This screw pin anchor shackle is forged, quenched, and tempered, featuring alloy pins for durability. It is fatigue-rated and designed for a wide range of temperatures, from -40° C to 204° C. The shackle meets or exceeds all requirements of ASME B30.26 and federal specifications, ensuring reliable performance in demanding applications. Each shackle is permanently marked with its working load limit and grade '6', signifying genuine quality.
  • Capacities from 1/3 through 55 metric tons.
  • Sizes 7/16 inch and above are hot dip galvanized.
  • Fatigue rated design ensures long-term performance.
  • Proof testing with certificates can be furnished upon request.
  • Can meet charpy requirements of 42 Joules average at -20° C upon special request.
  • Maximum proof load is 2 times the working load limit.
  • Minimum ultimate strength is 6 times the working load limit.