Learn More
MSC Crosby Screw Pin Anchor Shackle, 7/8 inch, Galvanized, 6.5 ton
Catalog No. 5000532892
Encompass
This screw pin anchor shackle combines the convenience of a quick-release pin with a strong, secure connection. Its bow shape supports a wider variety of connections, including multi-dimensional pulling, outperforming chain or D shackles. Made from forged, hot-dipped galvanized steel for corrosion resistance, it features an alloy pin for added strength. This high-performance shackle can meet DNV impact requirements of 42 Joules (31 ft-lb) at -20°C upon request, with proof-testing certificates available for various standards.
  • Working load limit, Grade 6, and PIC marked on each shackle.
  • Fatigue rated to 20,000 cycles at 1.5 times the WLL.
  • Meets Grade 6 shackle performance requirements.
  • Quick release screw pin for fast attachment and detachment, suitable for temporary lifting.
  • Anchor bow design, wider than D shackles, ideal for multi-leg rigging systems.
Catalog No. 50-005-32892 Supplier MSC Supplier No. 53601852
This screw pin anchor shackle combines the convenience of a quick-release pin with a strong, secure connection. Its bow shape supports a wider variety of connections, including multi-dimensional pulling, outperforming chain or D shackles. Made from forged, hot-dipped galvanized steel for corrosion resistance, it features an alloy pin for added strength. This high-performance shackle can meet DNV impact requirements of 42 Joules (31 ft-lb) at -20°C upon request, with proof-testing certificates available for various standards.
  • Working load limit, Grade 6, and PIC marked on each shackle.
  • Fatigue rated to 20,000 cycles at 1.5 times the WLL.
  • Meets Grade 6 shackle performance requirements.
  • Quick release screw pin for fast attachment and detachment, suitable for temporary lifting.
  • Anchor bow design, wider than D shackles, ideal for multi-leg rigging systems.