This alloy steel screw pin anchor shackle features a rounded shape, allowing it to handle loads from various directions without significant side load. It is suitable for multiple-leg slings and temporary applications requiring frequent connections and disconnections. The shackle is quenched and tempered for superior strength, ensuring reliable performance in demanding environments.
- Working load limit permanently displayed for safety.
- Hot dip galvanized finish for enhanced durability.
- Approved for use in temperatures from -40° C to 204° C.
- Meets or exceeds all ASME B30.26 requirements.
- Meets performance requirements of Federal Specification RR-C-271G.
- Can be supplied proof tested with certificates upon request.
- Features a red pin, indicating genuine quality.
- Material: forged alloy steel with alloy pins for strength.