Learn More
MSC Crosby Shackle, Screw Pin, 1500 lb Working Load Limit, Carbon Steel, 5/16 in Chain
Catalog No. 5000515247
Encompass
This Crosby shackle features a screw pin design and is constructed from durable carbon steel. It offers a 1500 lb working load limit, making it suitable for various lifting and rigging applications. This shackle is designed for use with 5/16 inch chains.
  • Screw pin design for secure fastening
  • 1500 lb working load limit for demanding tasks
  • Constructed from carbon steel for durability
  • Compatible with 5/16 inch nominal chain sizes
Catalog No. 50-005-15247 Supplier MSC Supplier No. 80834617
This Crosby shackle features a screw pin design and is constructed from durable carbon steel. It offers a 1500 lb working load limit, making it suitable for various lifting and rigging applications. This shackle is designed for use with 5/16 inch chains.
  • Screw pin design for secure fastening
  • 1500 lb working load limit for demanding tasks
  • Constructed from carbon steel for durability
  • Compatible with 5/16 inch nominal chain sizes