Learn More
MSC Crosby Wire Rope Lubricant, Vitalife 400, 12 oz, Aerosol Can
Catalog No. 5000529200
Encompass
This standard wire rope lubricant provides essential inner strand preservation and lubricity. It reduces friction and extends rope life, while adhering to strands to form a protective outer film against corrosion. The non-tacky formula prevents dust attraction and penetrates the rope to displace water and contaminants, minimizing wear.
  • Provides inner strand preservation and lubricity.
  • Allows for easy visual inspection of the ropes.
  • Reduces friction between wire rope strands, extending rope life.
  • Adheres to the surface of strands, forming an outer film that provides excellent corrosive protection.
  • Non-tacky, which prevents dust attraction.
  • Penetrates into wire rope to displace water and contaminants, reducing wear and corrosion.
Catalog No. 50-005-29200 Supplier MSC Supplier No. 20294286
This standard wire rope lubricant provides essential inner strand preservation and lubricity. It reduces friction and extends rope life, while adhering to strands to form a protective outer film against corrosion. The non-tacky formula prevents dust attraction and penetrates the rope to displace water and contaminants, minimizing wear.
  • Provides inner strand preservation and lubricity.
  • Allows for easy visual inspection of the ropes.
  • Reduces friction between wire rope strands, extending rope life.
  • Adheres to the surface of strands, forming an outer film that provides excellent corrosive protection.
  • Non-tacky, which prevents dust attraction.
  • Penetrates into wire rope to displace water and contaminants, reducing wear and corrosion.