Learn More
Medchemexpress LLC Croscarmellose sodium | 74811-65-7 | 50 MG
SDP
Catalog No. 5000401620
Encompass_Preferred
Croscarmellose sodium is a pharmaceutical additive approved by the FDA, functioning as an excipient. It acts as a suspending agent, binder, glidant, antiadherent, and disintegrant, improving the stability, solubility, and processability of pharmaceutical preparations and affecting co-administered drug ADME processes.
  • Approved by the US Food and Drug Administration (FDA).
  • Functions as a suspending agent, binder, glidant, antiadherent, and disintegrant.
  • Improves stability, solubility, and processability of pharmaceutical preparations.
  • Affects ADME processes of co-administered drugs.
  • Promotes dissolution of poorly soluble compounds in water.
  • Improves fluidity of drugs as a lubricant in tablets.
  • Prevents drug particles from adhering to molds.
  • Accelerates drug release and absorption in the gastrointestinal tract.
  • Used for thickening or stabilization in the food industry.
Catalog No. 50-004-01620 Supplier Medchemexpress LLC Supplier No. HY4507250MG
Croscarmellose sodium is a pharmaceutical additive approved by the FDA, functioning as an excipient. It acts as a suspending agent, binder, glidant, antiadherent, and disintegrant, improving the stability, solubility, and processability of pharmaceutical preparations and affecting co-administered drug ADME processes.
  • Approved by the US Food and Drug Administration (FDA).
  • Functions as a suspending agent, binder, glidant, antiadherent, and disintegrant.
  • Improves stability, solubility, and processability of pharmaceutical preparations.
  • Affects ADME processes of co-administered drugs.
  • Promotes dissolution of poorly soluble compounds in water.
  • Improves fluidity of drugs as a lubricant in tablets.
  • Prevents drug particles from adhering to molds.
  • Accelerates drug release and absorption in the gastrointestinal tract.
  • Used for thickening or stabilization in the food industry.