Learn More
Medchemexpress LLC Croscarmellose sodium | 74811-65-7 | 500 MG
SDP
Catalog No. 5000401954
Encompass_Preferred
Croscarmellose sodium is a pharmaceutical additive approved by the FDA, serving as an excipient to enhance the stability, solubility, and processability of pharmaceutical preparations. It plays a role in the dissolution of poorly soluble compounds, acts as a suspending agent, and improves drug fluidity in tablets. Additionally, it is utilized in the food industry for thickening or stabilization.
  • Functions as a suspending agent, binder, glidant, antiadherent, and disintegrant.
  • Improves stability, solubility, and processability of pharmaceutical preparations.
  • Promotes dissolution of poorly soluble compounds in water.
  • Acts as a lubricant in tablets to improve drug fluidity.
  • Prevents drug particles from adhering to molds.
  • Accelerates drug release and absorption in the gastrointestinal tract.
  • Used for thickening or stabilization in the food industry.
Catalog No. 50-004-01954 Supplier Medchemexpress LLC Supplier No. HY45072500MG
Croscarmellose sodium is a pharmaceutical additive approved by the FDA, serving as an excipient to enhance the stability, solubility, and processability of pharmaceutical preparations. It plays a role in the dissolution of poorly soluble compounds, acts as a suspending agent, and improves drug fluidity in tablets. Additionally, it is utilized in the food industry for thickening or stabilization.
  • Functions as a suspending agent, binder, glidant, antiadherent, and disintegrant.
  • Improves stability, solubility, and processability of pharmaceutical preparations.
  • Promotes dissolution of poorly soluble compounds in water.
  • Acts as a lubricant in tablets to improve drug fluidity.
  • Prevents drug particles from adhering to molds.
  • Accelerates drug release and absorption in the gastrointestinal tract.
  • Used for thickening or stabilization in the food industry.