Learn More
IDEX Health & Science Upchurch CROSS/.020 THRU HOLES SS
Catalog No. NC1919397
Encompass_Preferred
Idex High-Pressure Tees, Diameter Outer: 0.5 mm, Cross, Stainless Steel, Diameter Inner: 0.02 in., For Use With: 1/16 in. O.D. Tubing
Catalog No. NC1919397 Supplier IDEX Health & Science Upchurch Supplier No. U430
Idex High-Pressure Tees, Diameter Outer: 0.5 mm, Cross, Stainless Steel, Diameter Inner: 0.02 in., For Use With: 1/16 in. O.D. Tubing