Learn More
Roboz Surgical CROSS ACTION 38IN W/RATCHET AN
SDP
Catalog No. 5000574499
Encompass
This cross action retractor is a specialized surgical instrument designed for delicate tissue retraction, featuring a 3.8 in. total length and a convenient ratcheting mechanism to maintain tension during procedures. The instrument is constructed with sharp, angled prongs to ensure secure tissue engagement in confined surgical fields.
  • Equipped with a secure ratcheting mechanism for hands-free retraction
  • Features sharp, angled 4x4 prong design for precise tissue grasping
  • Provides a maximum spread of 25 mm
  • Includes 5 mm deep prongs for effective retraction in micro-surgical applications
  • Total length of 3.8 in. for optimal handling
Catalog No. 50-005-74499 Supplier Roboz Surgical Supplier No. RS6523
This cross action retractor is a specialized surgical instrument designed for delicate tissue retraction, featuring a 3.8 in. total length and a convenient ratcheting mechanism to maintain tension during procedures. The instrument is constructed with sharp, angled prongs to ensure secure tissue engagement in confined surgical fields.
  • Equipped with a secure ratcheting mechanism for hands-free retraction
  • Features sharp, angled 4x4 prong design for precise tissue grasping
  • Provides a maximum spread of 25 mm
  • Includes 5 mm deep prongs for effective retraction in micro-surgical applications
  • Total length of 3.8 in. for optimal handling