Learn More
Stoelting™ Cross Action Retractor with Ratchet
SDP
Catalog No. 10001416 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Angled
Straight
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
10-001-416 Angled
10-001-417 Straight
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-001-416 Supplier Stoelting™ Supplier No. 5212615
Add to Cart
Add to Cart
Stoelting™ Cross Action Retractor with Ratchet can be used to actively separate the edges of a surgical incision or wound, and can hold back underlying organs and tissues, so that body parts under the incision may be accessed.
  • Sharp retractors are better at providing a secure grip and they can safely hold back the skin, muscles, fascia, tendons, or the periosteum but they may damage vessels, nerves, the peritoneum, glands or cysts.
  • Blunt retractors are recommended when iatrogenic injury is a concern.
  • The main benefit of blunt retractors is that they prevent unintended injury to the tissue held by the retractor or to the underlying tissue.
Specifications
For Use With (Application) Neuroscience
Length (English) 4 in.
Model Angled
Includes Ratchet