Learn More
MSC Honeywell Miller Cross Arm Anchor Strap, 4 ft, Polyester, 2 D-Rings, 1 ea
Catalog No. 5000507121
Encompass
The cross arm anchor strap is a temporary anchorage connector designed for use within fall protection systems. Constructed from durable webbing, this anchor provides a reliable connection point for fall arrest and restraint applications.
  • Length of 4 ft
  • Includes one 2-in. D-ring and one 3-in. D-ring
  • Load capacity of 400 lbs.
  • Minimum tensile strength of 5,000 lbs.
  • Compliant with ANSI A10.32, ANSI Z359-2007, OSHA 1910.66, and OSHA 1926.502
Catalog No. 50-005-07121 Supplier MSC Supplier No. 91399170
The cross arm anchor strap is a temporary anchorage connector designed for use within fall protection systems. Constructed from durable webbing, this anchor provides a reliable connection point for fall arrest and restraint applications.
  • Length of 4 ft
  • Includes one 2-in. D-ring and one 3-in. D-ring
  • Load capacity of 400 lbs.
  • Minimum tensile strength of 5,000 lbs.
  • Compliant with ANSI A10.32, ANSI Z359-2007, OSHA 1910.66, and OSHA 1926.502