Learn More
Phenomenex Inc joins four 1/32"" OD tubing w/Fittings (MAX 15,000 psi / 1034 bar)
Catalog No. 502400991 Shop All Phenomenex Products
Encompass_Preferred
Cross Assembly, SS, 0.010" thruhole AN0-1040
Catalog No. 50-240-0991 Supplier Phenomenex Inc Supplier No. AN01040
Cross Assembly, SS, 0.010" thruhole AN0-1040