Learn More
IDEX Health & Science Upchurch Cross Assembly Tefzel (ETFE) for 1/16" OD
Catalog No. NC1600486
Encompass_Preferred
Our Tefzel (ETFE) Low Pressure Cross Assembly can handle a pressure rating 1000 psi (69 bar). It is designed with convenient mounting holes. The P-634 is designed for 1/16"OD tubing. This assembly comes complete with (4) P-245 Nuts and (4) P-200N Ferrules.
Catalog No. NC1600486 Supplier IDEX Health & Science Upchurch Supplier No. P634
Our Tefzel (ETFE) Low Pressure Cross Assembly can handle a pressure rating 1000 psi (69 bar). It is designed with convenient mounting holes. The P-634 is designed for 1/16"OD tubing. This assembly comes complete with (4) P-245 Nuts and (4) P-200N Ferrules.