Learn More
Chemglass Life Sciences Cross Connector with Male Luer Lock, L/S Platinum Cured Silicone Pump Tubing, 1/8" ID x 1/4" OD Size 16
SDP
Supplier:  Chemglass Life Sciences CLS139024
Encompass
Reduces set up time and minimizes labor cost. cross connector and male luer assembly are package 10 per bag and ready to use.Manufactured with 5" of No.16 (1/8 x 1/4") platinum cured silicone tubing.For sterile applications the assembly can be safely autoclaved.
Catalog No. 50-000-83287