Learn More
Electron Microscopy Sciences Cross Line Scale Reticle RET 21, 19 mm, 20 mm Scale, 0.10 mm Divisions
SDP
Catalog No. 501901553
Encompass_Preferred
High quality, precision made standard reticles are manufactured to use with our microscope eyepieces. All recticles are chrome on glass and positive polarity.
Size: 19 mm
Scale: 20 mm
Divisions: 0.10 mm
Catalog No. 50-190-1553 Supplier Electron Microscopy Sciences Supplier No. 6026719
High quality, precision made standard reticles are manufactured to use with our microscope eyepieces. All recticles are chrome on glass and positive polarity.
Size: 19 mm
Scale: 20 mm
Divisions: 0.10 mm