Learn More
Electron Microscopy Sciences Cross Line Scale Reticle RET 21, 21 mm, 20 mm Scale, 0.10 mm Divisions
SDP
Catalog No. 501901554
Encompass_Preferred
High quality, precision made standard reticles are manufactured to use with our microscope eyepieces. All recticles are chrome on glass and positive polarity.
Size: 21 mm
Scale: 20 mm
Divisions: 0.10 mm
Catalog No. 50-190-1554 Supplier Electron Microscopy Sciences Supplier No. 6026721
High quality, precision made standard reticles are manufactured to use with our microscope eyepieces. All recticles are chrome on glass and positive polarity.
Size: 21 mm
Scale: 20 mm
Divisions: 0.10 mm