Learn More
Grainger Cross Pein Hammer 2 lb 14-1/2 Hickory
Catalog No. 502765605
Encompass_Preferred
The Vaughan 16 oz. Cross Pein Hammer is a 16 ounce hammer with a cross pein head made of forged carbon steel for durability. It has a hickory handle and is designed for general purpose hammering and striking. This hammer is made in the USA and measures 13.5 inches in overall length. The cross pein head allows hammering with the flat face or rounded pein. It is ideal for metalworking, carpentry, masonry and other general hammering applications.
Catalog No. 50-276-5605 Supplier Grainger Supplier No. 16W197
The Vaughan 16 oz. Cross Pein Hammer is a 16 ounce hammer with a cross pein head made of forged carbon steel for durability. It has a hickory handle and is designed for general purpose hammering and striking. This hammer is made in the USA and measures 13.5 inches in overall length. The cross pein head allows hammering with the flat face or rounded pein. It is ideal for metalworking, carpentry, masonry and other general hammering applications.