Learn More
IDEX Health & Science Upchurch CROSS PK .020THRU HOLE W/F-300
Catalog No. NC1919176
Encompass_Preferred
Idex High-Pressure Crosses, Pressure Rating: 3500 psi, Black, Diameter Inner: 0.02 in., For Use With: 1/16 in. O.D. Tubing, PEEK, Swept Volume: 0.721 uL.
Catalog No. NC1919176 Supplier IDEX Health & Science Upchurch Supplier No. P729
Idex High-Pressure Crosses, Pressure Rating: 3500 psi, Black, Diameter Inner: 0.02 in., For Use With: 1/16 in. O.D. Tubing, PEEK, Swept Volume: 0.721 uL.