Learn More
Description
Includes
Activity and fact guide
- Examine the layers of the Earth with this brightly colored, 5 in. dia. (12.5cm) foam model
- Layers separate to reveal planet core
Specifications
Specifications
| For Use With (Application) | Illustrates both continents and oceans with raised features |
| Product Type | Cross Section Earth Model |
| Includes | Activity and fact guide |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
