Learn More
Electron Microscopy Sciences Cross Sectional Holder
SDP
Catalog No. 5025160
Encompass
Made from non-magnetic stainless steel with 3.1 mm (1/8") diameter pin and adjustable angle turn-screw. Just insert specimens edge-on and observe the cross section directly.
Catalog No. 50-251-60 Supplier Electron Microscopy Sciences Supplier No. 7594201
Made from non-magnetic stainless steel with 3.1 mm (1/8") diameter pin and adjustable angle turn-screw. Just insert specimens edge-on and observe the cross section directly.