Learn More
MSC CrossFire Magnifying Safety Glasses, 2.00 Diopter, ANSI Z87.1, 1 ea
Catalog No. 5000770119
Encompass
These safety glasses are designed with magnifying lenses to enhance visibility for detailed tasks. Featuring a full-framed, wrap-around design, they provide protection and comfort in various work environments.
  • Magnifying diopter lens (2.00) for enhanced vision
  • Anti-fog and scratch-resistant lens coatings
  • Clear polycarbonate lenses with UV protection
  • Full-framed, wrap-around design with black frame and temples
  • Meets ANSI Z87.1 safety standards
  • Foam-lined for added comfort and protection
  • Non-adjustable temples for a secure fit
Catalog No. 50-007-70119 Supplier MSC Supplier No. 44318541
These safety glasses are designed with magnifying lenses to enhance visibility for detailed tasks. Featuring a full-framed, wrap-around design, they provide protection and comfort in various work environments.
  • Magnifying diopter lens (2.00) for enhanced vision
  • Anti-fog and scratch-resistant lens coatings
  • Clear polycarbonate lenses with UV protection
  • Full-framed, wrap-around design with black frame and temples
  • Meets ANSI Z87.1 safety standards
  • Foam-lined for added comfort and protection
  • Non-adjustable temples for a secure fit