Learn More
MSC CrossFire Safety Glasses, Anti-Fog, Clear Lenses, Full-Framed, 1 ea
Catalog No. 5000787924
Encompass
These safety glasses feature a full-framed design with clear polycarbonate lenses and an anti-fog coating to enhance visibility in hot and humid conditions. They comply with ANSI Z87.1+ standards.
  • Anti-fog lens coating
  • Full-framed style
  • Wrap around frame design
  • Clear lens color
  • Black frame color
  • Universal size
  • Polycarbonate lens material
  • Non-adjustable temples
  • UV protection
  • Foam lined
Catalog No. 50-007-87924 Supplier MSC Supplier No. 92483932
These safety glasses feature a full-framed design with clear polycarbonate lenses and an anti-fog coating to enhance visibility in hot and humid conditions. They comply with ANSI Z87.1+ standards.
  • Anti-fog lens coating
  • Full-framed style
  • Wrap around frame design
  • Clear lens color
  • Black frame color
  • Universal size
  • Polycarbonate lens material
  • Non-adjustable temples
  • UV protection
  • Foam lined