Learn More
Thermo Scientific™ Crossflow Filtration Accessory Pack
Catalog No. FERA57937
Change view
Click to view available options
Quantity:
Each
Catalog No. Quantity
FERA57937 Each
1 options
Catalog No. FERA57937 Supplier Thermo Scientific™ Supplier No. A57937
Add to Cart
Edge
Add to Cart
The Crossflow Filtration Accessory Pack features all necessary components including the feed line, return line, filtrate line, and pressure gauge.
Accessory pack includes:
• Feed line
• Return line
• Filtrate line
• Pressure gauge
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Not Autoclavable
Disposable Reusable
For Use With (Equipment) Filtration Systems
Material PVC, Polypropylene, Stainless Steel
Sterility Non-sterile
Includes Feed line, return line, filtrate line, and pressure gauge
Quantity Each
Fit for Purpose Certified Quality