Learn More
BioVendor, LLC CROSSLAP CULTURE CTX-I EIA (96 wells)
SDP
Supplier:  BioVendor, LLC AC07F1
Encompass
Sandwich ELISA, HRP-labelled antibodyThe test is an enzyme-linked immunosorbent assay (ELISA) for the quantitative determination of degradation products of C-terminal telopeptides of type I collagen in bone cell culture supernatants.Type I collagen accounts for more than 90% of the organic matrix of bone. The resorption process can be studied in vitro by culturing bone cells on devitalised slices of bone (catalog # DT-1BON1000-96) or dentin. ApplicationsCell culture supernatantSample Requirements50 μL Pre-diluted (1:5)Calibration Range0-112.7 nmLimit of Detection0.44 nM CrossLaps
Catalog No. 50-196-3838