Learn More
Med Vet International Crossley Molar Luxator, J0041MR
Catalog No. 5000584683
Encompass_Preferred
Designed to facilitate extraction of molars with a minimum of damage to the alveolus and surrounding soft tissue. One cutting blade is for insertion down buccal or lingual surface. The other end of the instrument is set at right angles to penetrate between molars.
Catalog No. 50-005-84683 Supplier Med Vet International Supplier No. JOR41MR
Designed to facilitate extraction of molars with a minimum of damage to the alveolus and surrounding soft tissue. One cutting blade is for insertion down buccal or lingual surface. The other end of the instrument is set at right angles to penetrate between molars.