Learn More
Med Vet International Crossley Rabbit Incisor Luxator, J0041CR
Catalog No. 5000585332
Encompass_Preferred
Incisor malocclusion with resulting over growth is a common and serious problem in rabbits. Frequent clipping is required with the risk of splitting the incisors. Extraction is the only permanent solution but the roots of the incisors are large relative to the jaw. The Crossley incisor luxatorwas designed especially for this procedure. The instrument matches the shape of the rabbit incisor. It is double ended for various size incisors. Complete with instructions for use.
Catalog No. 50-005-85332 Supplier Med Vet International Supplier No. JOR41CR
Incisor malocclusion with resulting over growth is a common and serious problem in rabbits. Frequent clipping is required with the risk of splitting the incisors. Extraction is the only permanent solution but the roots of the incisors are large relative to the jaw. The Crossley incisor luxatorwas designed especially for this procedure. The instrument matches the shape of the rabbit incisor. It is double ended for various size incisors. Complete with instructions for use.