Learn More
Electron Microscopy Sciences Cross Scale Reticle RET 30, 21 mm, 10 mm Scale, 0.10 mm x 100 Divisions
SDP
Catalog No. 501901755
Encompass_Preferred
High quality, precision made standard reticles are manufactured to use with our microscope eyepieces. All recticles are chrome on glass and positive polarity.
Size: 21 mm
Scale: 10 mm
Divisions: 0.10 mm x 100
Catalog No. 50-190-1755 Supplier Electron Microscopy Sciences Supplier No. 6027821
High quality, precision made standard reticles are manufactured to use with our microscope eyepieces. All recticles are chrome on glass and positive polarity.
Size: 21 mm
Scale: 10 mm
Divisions: 0.10 mm x 100