Learn More
Electron Microscopy Sciences Cross Scale Reticle RET 32, 19 mm, 20 mm Scale, 0.10 mm x 200 Divisions
SDP
Catalog No. 501901756
Encompass_Preferred
High quality, precision made standard reticles are manufactured to use with our microscope eyepieces. All recticles are chrome on glass and positive polarity.
Size: 19 mm
Scale: 20 mm
Divisions: 0.10 mm x 200
Catalog No. 50-190-1756 Supplier Electron Microscopy Sciences Supplier No. 6027919
High quality, precision made standard reticles are manufactured to use with our microscope eyepieces. All recticles are chrome on glass and positive polarity.
Size: 19 mm
Scale: 20 mm
Divisions: 0.10 mm x 200