Learn More
Cole-Parmer Crosstex Self-Seal Sterilization Pouches, Standard, 12" x 18"; 100/Box, 500/Case
Catalog No. NC1885150 Shop All Cole-Parmer Products
Encompass_Preferred
Provides breathability, a sterile barrier, and indication of processing without the need to heat seal. Perfect for small instruments that require frequent sterilization. Multi-parameter pouches confirm all sterilization criteria time, temperature, and steam.
Catalog No. NC1885150 Supplier Cole-Parmer Supplier No. 1070349
Provides breathability, a sterile barrier, and indication of processing without the need to heat seal. Perfect for small instruments that require frequent sterilization. Multi-parameter pouches confirm all sterilization criteria time, temperature, and steam.