Learn More
Enzo Life Sciences Crosstide (biotinylated) (1mg)
SDP
Supplier:  Enzo Life Sciences BMLP1910001
Encompass_Preferred
This peptide sequence, biotin-GRPRTSSFAEG, corresponds to the sequence of GSK3 surrounding the serine phosphorylated by MAP kinase activated protein kinase (MAPKAP kinase-1) and p70 ribosomal S6 kinase. Acts as a substrate for Akt (PKB), Km=4 µM. The N-terminal biotin allows this peptide to be used in kinase assays with streptavidin-bound membranes. Purity: ≥95%. Solubility: Soluble in Water. Long Term Storage: -20°C.
Catalog No. 50-201-1272