Learn More
Aviva Systems Biology Corp CROT Antibody - N-terminal region (ARP48873_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48873_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CROT. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CROT. Alternate names: COTApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-59945