Learn More
Medchemexpress LLC Crotamine TFA | 98.06% | 4883.73 (free base) | 1 MG
SDP
Catalog No. 5000398397
Encompass_Preferred
Crotamine TFA is a Na+ channel modulator, a 42 amino acid toxin cross-linked by three disulfide bridges. It exhibits analgesic activity and interacts with lipid membranes, showing myonecrotic activity. It can be isolated from *Crotalus durissus terrificus* venom.
  • Na+ channel modulator
  • 42 amino acid toxin
  • Cross-linked by three disulfide bridges
  • Analgesic activity
  • Interacts with lipid membranes, showing myonecrotic activity
Catalog No. 50-003-98397 Supplier Medchemexpress LLC Supplier No. HYP5159A1MG
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Crotamine TFA is a Na+ channel modulator, a 42 amino acid toxin cross-linked by three disulfide bridges. It exhibits analgesic activity and interacts with lipid membranes, showing myonecrotic activity. It can be isolated from *Crotalus durissus terrificus* venom.
  • Na+ channel modulator
  • 42 amino acid toxin
  • Cross-linked by three disulfide bridges
  • Analgesic activity
  • Interacts with lipid membranes, showing myonecrotic activity