Learn More
Medchemexpress LLC Crotamine (TFA) | 98.1% | 4883.73 | 5 MG
SDP
Catalog No. 5000398504
Encompass_Preferred
Crotamine TFA is a Na+ channel modulator. It is a 42 amino acid toxin cross-linked by three disulfide bridges. It has analgesic activity and also interacts with lipid membranes, showing myonecrotic activity. Crotamine can be isolated from *Crotalus durissus terrificus* venom.
  • Na+ channel modulator
  • 42 amino acid toxin
  • Cross-linked by three disulfide bridges
  • Analgesic activity
  • Interacts with lipid membranes
  • Shows myonecrotic activity
  • Isolated from *Crotalus durissus terrificus* venom
Catalog No. 50-003-98504 Supplier Medchemexpress LLC Supplier No. HYP5159A5MG
Crotamine TFA is a Na+ channel modulator. It is a 42 amino acid toxin cross-linked by three disulfide bridges. It has analgesic activity and also interacts with lipid membranes, showing myonecrotic activity. Crotamine can be isolated from *Crotalus durissus terrificus* venom.
  • Na+ channel modulator
  • 42 amino acid toxin
  • Cross-linked by three disulfide bridges
  • Analgesic activity
  • Interacts with lipid membranes
  • Shows myonecrotic activity
  • Isolated from *Crotalus durissus terrificus* venom