Learn More
Enzo Life Sciences Croton oil (2.25kg). CAS: 8001-28-3
SDP
Supplier:  Enzo Life Sciences ALX350089KI225
Encompass_Preferred
Natural source of phorbol and phorbol esters, purified from Croton tiglium L. Alternative name: Tiglium oil. Appearance: Pale-yellow to brown-yellow oil. Solubility: Soluble in DMSO, 100% ethanol or acetone. Insoluble in cold water. Long Term Storage: +4°C.
Catalog No. 50-200-8726