Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Croton Oil, Spectrum™ Chemical
SDP

Catalog No. 18601888 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
1 kg
12 kg
Packaging:
Amber Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-888 12 kg Amber Glass Bottle
18-601-883 10 g Amber Glass Bottle
18-601-884 25 g Amber Glass Bottle
18-601-885 100 g Amber Glass Bottle
18-601-886 500 g Amber Glass Bottle
18-601-887 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-601-888 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. CR13212KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

CR132, 8001-28-3

Spectrum™ Chemical Croton Oil is an oil prepared from the seeds of the tree, Croton tiglium. Croton oil is the source of the organic compound phorbol and is the basis of rejuvenating chemical peels, due to the caustic exfoliating effects it has on the dermal components of the skin. Used in conjunction with phenol solutions, it results in an intense reaction which leads to initial skin sloughing and then eventual regeneration. In traditional Chinese medicine it is used as an ingredient in some liniments.

Chemical Identifiers

CAS 8001-28-3

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Refractive Index 1.4760-1.4780
Quantity 12 kg
UN Number UN2810
Grade Ungraded
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material Croton Oil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.