Learn More
Medchemexpress LLC Crotonoside | 1818-71-9 | 99.8% | 283.24 g/mol | C10H13N5O5 | 10 MG
SDP
Catalog No. 5000296882
Encompass_Preferred
Crotonoside (isoguanosine) is a natural nucleoside isolated from Croton species that has reported inhibitory activity against FLT3 and HDAC3/6 and selective activity in acute myeloid leukemia (AML) cells. Supplied for research use only.
  • Natural nucleoside with reported FLT3 and HDAC3/6 inhibitory activity.
  • Suitable for preclinical biochemical and cellular studies.
  • High purity, supported by batch certificate of analysis.
  • Molecular formula C10H13N5O5; molecular weight 283.24 g/mol.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-96882 Supplier Medchemexpress LLC Supplier No. HYN007110MG
Crotonoside (isoguanosine) is a natural nucleoside isolated from Croton species that has reported inhibitory activity against FLT3 and HDAC3/6 and selective activity in acute myeloid leukemia (AML) cells. Supplied for research use only.
  • Natural nucleoside with reported FLT3 and HDAC3/6 inhibitory activity.
  • Suitable for preclinical biochemical and cellular studies.
  • High purity, supported by batch certificate of analysis.
  • Molecular formula C10H13N5O5; molecular weight 283.24 g/mol.
  • Intended for research use only; not for human or clinical use.