Learn More
Medchemexpress LLC HY-N0071 5mg Medchemexpress, Crotonoside CAS:1818-71-9 Purity:>98%
SDP
Catalog No. 501970219
Encompass_Preferred
Medchemexpress, HY-N0071 5mg Crotonoside CAS:1818-71-9 Crotonoside is isolated from Chinese medicinal herb, Croton. Crotonoside inhibits FLT3 and HDAC3/6, exhibits selective inhibition in acute myeloid leukemia (AML) cells. Crotonoside could be a promising new lead compound for the treatment of AML. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0219 Supplier Medchemexpress LLC Supplier No. HYN00715MG
Medchemexpress, HY-N0071 5mg Crotonoside CAS:1818-71-9 Crotonoside is isolated from Chinese medicinal herb, Croton. Crotonoside inhibits FLT3 and HDAC3/6, exhibits selective inhibition in acute myeloid leukemia (AML) cells. Crotonoside could be a promising new lead compound for the treatment of AML. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.