Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Crotyl alcohol, mixture of cis and trans

Catalog No. C86006100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C86006100ML 100 mL
C8600625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C86006100ML Supplier Sigma Aldrich Supplier No. C86006100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Crotyl alcohol can be used as a starting material in the synthesis of antitumor agents such as 14-azacamptothecin and 10, 11-methylenedioxy-14-azacamptothecin
  • It can also be used as a precursor in the total synthesis of discodermolide

Specifications

CAS 6117-91-5
Density 0.845 g/mL (at 25°C (literature))
Boiling Point 121°C to 122°C (lit.)
Molecular Formula C4H8O
Refractive Index n20/D 1.427 (literature)
Linear Formula CH3CH=CHCH2OH
MDL Number MFCD00002923
Quantity 100 mL
Synonym 2-Buten-1-ol
Molecular Weight (g/mol) 72.11
Percent Purity 96%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.