Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Crotyl bromide

Catalog No. C864055G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C864055G 5 g
C8640525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C864055G Supplier Sigma Aldrich Supplier No. C864055G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Crotyl bromide is a general organic reagent that can be used in the total synthesis of biologically important natural products and their derivatives such as (+)-artemisinin, plakortone E and ()-horsfiline.It can also be used to prepare analogs of deoxycyclitols, pentopyranoses, 6-deoxyhexoses, hexoses and pseudo-indoxyl derivatives.remainder 3-bromo-1-butene

Specifications

CAS 29576-14-5
Density 1.312 g/mL (at 25°C (literature))
Boiling Point 97°C to 99°C (lit.)
Molecular Formula C4H7Br
Refractive Index n20/D 1.480 (literature)
Linear Formula CH3CH=CHCH2Br
MDL Number MFCD00000248
Quantity 5 g
Synonym trans-1-Bromo-2-butene
Molecular Weight (g/mol) 135
Percent Purity 85%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.