Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Crotyl chloride, predominantly trans

Catalog No. 254584100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-458-4100G 100 g
25-458-425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 25-458-4100G Supplier Sigma Aldrich Supplier No. 254584100G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Crotyl chloride reacts with trichlorosilane in the presence of tertiary amine and copper salt to give the corresponding allyltrichlorosilane.crotyl chloride was used in kinetics measurements of reactions of atomic chlorine with series of unsaturated aldehydes and ketones at 298k
  • (e)crotyl chloride was also used in the preparation of (e)-crotyltrichlorosilane

Specifications

CAS 4894-61-5
Density 0.929 g/mL (at 25°C (literature))
Boiling Point 84°C to 85°C (lit.)
Molecular Formula C4H7Cl
Refractive Index n20/D 1.436 (literature)
Linear Formula CH3CH=CHCH2Cl
Quantity 100 g
MDL Number MFCD00000987
Synonym 1-Chloro-2-butene
Molecular Weight (g/mol) 90.55
Percent Purity 95%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.